What is IRMS?
Isotope ratio mass spectrometry (IRMS) is the study of natural and synthetic samples based on their isotope ratios. The isotope ratio of a material will vary depending on its source and origin as well as on processes that may occur during the lifetime of that material. While IRMS is widely used in Earth sciences to understand Earth’s geological history, it is also used in food authenticity, forensics, medical research, and antidoping testing.
We offer a comprehensive portfolio of isotope ratio mass spectrometers. With more than 70 years of experience building cutting edge analytic instrumentation and with frequent new innovations across the portfolio, our isotope ratio mass spectrometers are the obvious choice for your analytical needs.
Noble gas mass spectrometry
Select from our portfolio of noble gas mass spectrometers to obtain reliable analytical results in the fields of dating, geochronology, and thermochronology.
Learn more about noble gas MS ›
New dimensions in isotope ratio analysis
Orbitrap Exploris Isotope Solutions
Orbitrap Exploris Isotope Solutions
Open new dimensions in isotope ratio analysis with Thermo Scientific Orbitrap Exploris Isotope Solutions. Preserve intramolecular isotopic information throughout the analysis and determine previously inaccessible isotopologues by the combination of soft electrospray ionization (ESI) and high-resolution accurate mass (HRAM) sensitivity.
Visit product page  Download brochure  Watch video
Retarding Potential Quadrupole (RPQ)
The RPQ improves abundance sensitivity by an order of magnitude and acts as a high selectivity energy filter for ions with disturbed energy or angle. Both our MC-ICP-MS and TIMS can be configured with two RPQ lenses.
Learn about RPQ option ›
Amplify your precision with 1013 amplifier technology
Limited sample size? Equipped with 1013 ohm amplifiers, our spectrometers enable faster response times and extremely low noise/signal characteristics.
Learn about 1013 Ohm Amplifier Technology ›
Add MS/MS to your Thermo Scientific Neoma MC-ICP-MS
Neoma MC-ICP-MS comes with the upgrade pathway to MS/MS MC-ICP-MS. This dedicated collision/reaction cell MC-ICP-MS, with unique pre-cell mass filtering technology, allows separation of isobaric interferences.
Learn about adding MS/MS to your Neoma MC-ICP-MS ›
Qtegra ISDS Software for Gas IRMS
Automate your research by managing instrument control, data acquisition, data processing, and report generation from one easy software platform with Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS). Utilize this software with gas IRMS instruments, high resolution IRMS, and noble gas mass spectrometers. 
Learn more about Qtegra ISDS ›
Understanding the world around us and our place within it using isotopes.
From demise of the dinosaurs through to finding new ways to diagnose diseases, isotope research provides critical information about biological, chemical and geological processes. Isotopes can be used to provide information on how ancient civilizations lived, they can be used to date critical events in the geological past and can provide insights into disease pathways. Explore the ways that isotopes can benefit your field of research.
Archaeology
Geochronology
Battery
Nuclear
Biology & Biogeochemistry
Oil & Gas
Forensics
Semiconductor and high purity metals
* Required field
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes